missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 11486870
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73852-17-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00064869
InChI Key CXDPUSMFYPQXCV-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid
PubChem CID 2734332
IUPAC Name (2,6-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 151°C to 152°C
Quantity 1 g
Beilstein 4802506
Solubility Information Insoluble in water.
Formula Weight 190.82
Percent Purity 95%
Chemical Name or Material 2,6-Dichlorobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'