missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11437497
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11437497

Brand: Thermo Scientific Alfa Aesar A15615.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Dichlorobenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store in cool dry place in tightly closed container. Store under dry inert gas. Keep away from water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6579-54-0
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00052311
InChI Key WGGKQIKICKLWGN-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzene-1-sulfonyl chloride, 2,6-dichlorobenzenesulfonylchloride, 2,6-dichlorophenylsulfonyl chloride, 2,6-dichlorobenzenesulphonyl chloride, unii-8523kf52zn, benzenesulfonyl chloride, 2,6-dichloro, 2,6-dichloro-benzenesulfonyl chloride, 2,6-dichlorobenzene sulfonyl chloride, pubchem5117, acmc-209ns8
PubChem CID 522183
IUPAC Name 2,6-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)S(=O)(=O)Cl)Cl

Specifications

Melting Point 51°C to 55°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 1531023
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 245.51
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorobenzenesulfonyl chloride
compliance-icons
Product Identifier
  • 2,6-Dichlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.