Learn More
2,6-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals
Description
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.
Solubility
Soluble in water (0.018g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 143°C to 147°C |
Boiling Point | 270°C |
Odor | Aromatic |
Quantity | 10 g |
UN Number | UN3077 |
Beilstein | 1909167 |
Merck Index | 14,3042 |
Formula Weight | 172.01 |
Percent Purity | 98% |
Chemical Name or Material | 2,6-Dichlorobenzonitrile |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10249564 | Certificate of Analysis | 06/02/2024 | A14986.09,A14986.22,A14986.14 |
10236489 | Certificate of Analysis | 22/11/2021 | A14986.09,A14986.22,A14986.14 |
USLF005585 | Certificate of Analysis | 22/11/2021 | A14986.09,A14986.22,A14986.14 |
10226859 | Certificate of Analysis | 22/11/2021 | A14986.09,A14986.22,A14986.14 |
10236490 | Certificate of Analysis | 22/11/2021 | A14986.09,A14986.22,A14986.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.