missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals

23.72 € - 517.58 €

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key CVSUAFOWIXUYQA-UHFFFAOYSA-M
Synonym Tillman's reagent hydrate
PubChem CID 23696612
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10051830
View Documents
Thermo Scientific Acros
152870010
1 g Glass bottle
23.72 €
1g
In Stock
to see stock.
Add to basket
 
10636932
View Documents
Thermo Scientific Acros
152870100
10 g Glass bottle
86.63 €
10g
In Stock
to see stock.
Add to basket
 
10746161
View Documents
Thermo Scientific Acros
152870250
25 g Glass bottle
182.91 €
25g
In Stock
to see stock.
Add to basket
 
11985591
View Documents
Thermo Scientific Acros
152871000
100 g Glass bottle
517.58 €
100g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1266615-56-8
290.07
CVSUAFOWIXUYQA-UHFFFAOYSA-M
23696612
C12H6Cl2NNaO2
MFCD00150014
Tillman's reagent hydrate
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1

Specifications

1266615-56-8
C12H6Cl2NNaO2
Tillman's reagent hydrate
CVSUAFOWIXUYQA-UHFFFAOYSA-M
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
290.07
290.07
≥98%
Dark Green
2, 6-Dichloroindophenol, sodium salt hydrate
98+%
MFCD00150014
Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Authentic
sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
23696612
14% max. (1 g, 120°C)
For determination of ascorbic acid: Passes Test
Powder
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.