missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals

Product Code. 11437037
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-65-0
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002176
InChI Key HOLHYSJJBXSLMV-UHFFFAOYSA-N
Synonym phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp
PubChem CID 6899
ChEBI CHEBI:28457
IUPAC Name 2,6-dichlorophenol
SMILES OC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 65°C to 68°C
Boiling Point 218°C to 220°C
Odor Phenol-like
Quantity 25 g
UN Number UN2020
Beilstein 1447806
Merck Index 14,3073
Formula Weight 163
Percent Purity 99%
Chemical Name or Material 2,6-Dichlorophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'