missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyrazine, 98%, Thermo Scientific Chemicals

Product Code. 11424310
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,6-Dichloropyrazine is used in the preparation of pyrazine and pyridine compounds as ATR kinase inhibitors. It is also used in the synthesis of oxacalixarenes containing nitrogen heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloropyrazine is used in the preparation of pyrazine and pyridine compounds as ATR kinase inhibitors. It is also used in the synthesis of oxacalixarenes containing nitrogen heterocycles.

Solubility
Soluble in ethanol and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4774-14-5
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006125
InChI Key LSEAAPGIZCDEEH-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine
PubChem CID 78504
IUPAC Name 2,6-dichloropyrazine
SMILES C1=C(N=C(C=N1)Cl)Cl

Specifications

Melting Point 52°C to 56°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2439
Solubility Information Soluble in ethanol and methanol.
Formula Weight 148.98
Percent Purity 99%
Chemical Name or Material 2,6-Dichloropyrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.