Learn More
2,6-Difluoro-3-methoxyphenol, 97%, Thermo Scientific™
104.00 € - 359.00 €
Chemical Identifiers
CAS | 886498-60-8 |
---|---|
Molecular Formula | C7H6F2O2 |
Molecular Weight (g/mol) | 160.12 |
MDL Number | MFCD04115907 |
InChI Key | UOVAWRHEABQHGR-UHFFFAOYSA-N |
Synonym | 2,6-difluoro-3-methoxy-phenol, phenol,2,6-difluoro-3-methoxy, 2,6-bis fluoranyl-3-methoxy-phenol |
PubChem CID | 3682862 |
IUPAC Name | 2,6-difluoro-3-methoxyphenol |
SMILES | COC1=C(C(=C(C=C1)F)O)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
886498-60-8 | |
160.12 | |
UOVAWRHEABQHGR-UHFFFAOYSA-N | |
3682862 | |
COC1=C(C(=C(C=C1)F)O)F |
C7H6F2O2 | |
MFCD04115907 | |
2,6-difluoro-3-methoxy-phenol, phenol,2,6-difluoro-3-methoxy, 2,6-bis fluoranyl-3-methoxy-phenol | |
2,6-difluoro-3-methoxyphenol |
Specifications
886498-60-8 | |
C7H6F2O2 | |
2,6-difluoro-3-methoxy-phenol, phenol,2,6-difluoro-3-methoxy, 2,6-bis fluoranyl-3-methoxy-phenol | |
COC1=C(C(=C(C=C1)F)O)F | |
160.12 | |
160.12 | |
2,6-Difluoro-3-methoxyphenol |
73°C to 76°C | |
MFCD04115907 | |
UOVAWRHEABQHGR-UHFFFAOYSA-N | |
2,6-difluoro-3-methoxyphenol | |
3682862 | |
97% |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures