missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylaniline, 90+%, Thermo Scientific Chemicals

Product Code. 10206080
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24544-04-5
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.29
MDL Number MFCD00008887
InChI Key WKBALTUBRZPIPZ-UHFFFAOYSA-N
Synonym 2,6-diisopropylaniline, benzenamine, 2,6-bis 1-methylethyl, 2,6-diisopropyl aniline, aniline, 2,6-diisopropyl, diisopropylaniline, 2,6-diisopropylphenylamine, 2,6-bis propan-2-yl aniline, 2,6-dilsopropylaniline, 2,6-di propan-2-yl aniline, 2,6-bis 1-methylethyl aniline
PubChem CID 32484
IUPAC Name 2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point -45.0°C
Density 0.9400g/mL
Boiling Point 257.0°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CH]2C6H3NH2
Refractive Index 1.5320 to 1.5340
Quantity 500 g
Beilstein 12, 168
Specific Gravity 0.94
Solubility Information Solubility in water: <0.20g/L. Other solubilities: miscible with ethanol and toluene
Formula Weight 177.29
Percent Purity 90+%
Physical Form Liquid
Chemical Name or Material 2, 6-Diisopropylaniline, 92%
Show More Show Less
Product Identifier
  • 2,6-Diisopropylaniline
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.