Learn More
2,6-Dimethoxybenzeneboronic acid, 97+%, Thermo Scientific Chemicals
32.10 € - 166.83 €
Chemical Identifiers
CAS | 23112-96-1 |
---|---|
Molecular Formula | C8H11BO4 |
Molecular Weight (g/mol) | 181.982 |
MDL Number | MFCD01318987 |
InChI Key | BKWVXPCYDRURMK-UHFFFAOYSA-N |
Synonym | 2,6-dimethoxyphenyl boronic acid, 2,6-dimethoxyphenylboronicacid, 2,6-dimethoxybenzeneboronic acid, 2,6-dimethoxyphenyl-boronic acid, 2,6-dimethoxyphenyl boranediol, boronic acid, 2,6-dimethoxyphenyl, pubchem1825, acmc-1clhv, ksc201q0f, 2,6-dimethoxy-phenylboronic acid |
PubChem CID | 2734343 |
IUPAC Name | (2,6-dimethoxyphenyl)boronic acid |
SMILES | B(C1=C(C=CC=C1OC)OC)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445727
|
Thermo Scientific Alfa Aesar
B24305.03 |
1 g | N/A | ||||||
11455727
|
Thermo Scientific Alfa Aesar
B24305.06 |
5 g |
49.85 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465727
|
Thermo Scientific Alfa Aesar
B24305.14 |
25 g |
166.83 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23112-96-1 | |
181.982 | |
BKWVXPCYDRURMK-UHFFFAOYSA-N | |
2734343 | |
B(C1=C(C=CC=C1OC)OC)(O)O |
C8H11BO4 | |
MFCD01318987 | |
2,6-dimethoxyphenyl boronic acid, 2,6-dimethoxyphenylboronicacid, 2,6-dimethoxybenzeneboronic acid, 2,6-dimethoxyphenyl-boronic acid, 2,6-dimethoxyphenyl boranediol, boronic acid, 2,6-dimethoxyphenyl, pubchem1825, acmc-1clhv, ksc201q0f, 2,6-dimethoxy-phenylboronic acid | |
(2,6-dimethoxyphenyl)boronic acid |
Specifications
23112-96-1 | |
C8H11BO4 | |
3032352 | |
BKWVXPCYDRURMK-UHFFFAOYSA-N | |
(2,6-dimethoxyphenyl)boronic acid | |
2734343 | |
≥97% |
106°C to 108°C | |
MFCD01318987 | |
2,6-dimethoxyphenyl boronic acid, 2,6-dimethoxyphenylboronicacid, 2,6-dimethoxybenzeneboronic acid, 2,6-dimethoxyphenyl-boronic acid, 2,6-dimethoxyphenyl boranediol, boronic acid, 2,6-dimethoxyphenyl, pubchem1825, acmc-1clhv, ksc201q0f, 2,6-dimethoxy-phenylboronic acid | |
B(C1=C(C=CC=C1OC)OC)(O)O | |
181.982 | |
181.98 | |
2,6-Dimethoxybenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.