missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Product Code. 11355659
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221006-70-8
Molecular Formula C7H10BNO4
Molecular Weight (g/mol) 182.97
MDL Number MFCD03788239
InChI Key ADGHSWFUZUADDH-UHFFFAOYSA-N
Synonym 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid
PubChem CID 2762707
IUPAC Name (2,6-dimethoxypyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)OC)OC)(O)O

Specifications

Melting Point 128°C to 130°C
Quantity 250 mg
Beilstein 9391039
Formula Weight 182.97
Percent Purity 95%
Chemical Name or Material 2,6-Dimethoxypyridine-3-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'