missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dimethylthiophenol, 97%, Thermo Scientific Chemicals Product Code.: 11440853

2,6-Dimethylthiophenol, 97%, Thermo Scientific Chemicals

Product Code. 11440853
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11440853

Brand: Thermo Scientific Alfa Aesar A13286.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-72-9
Molecular Formula C8H9S
Molecular Weight (g/mol) 137.22
MDL Number MFCD00010021
InChI Key QCLJODDRBGKIRW-UHFFFAOYSA-M
Synonym 2,6-dimethylthiophenol, 2,6-dimethyl thiophenol, benzenethiol, 2,6-dimethyl, 2,6-xylenethiol, 2,6-xylyl mercaptan, 2,6-dimethylphenylthiol, 2,6-dimethyl-benzenethiol, fema no. 3666, 2,6-dimethylbenzolthiol, 2,6-thioxylenol
PubChem CID 61045
IUPAC Name 2,6-dimethylbenzenethiol
SMILES CC1=CC=CC(C)=C1[S-]

Specifications

Density 1.038
Boiling Point 122°C (50 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.5749
Quantity 5 g
UN Number UN2810
Beilstein 1099405
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylthiophenol
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethylthiophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Dimethylthiophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dimethylthiophenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.