missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11430063
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from strong acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-22-4
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.123
MDL Number MFCD00007148
InChI Key QFUSCYRJMXLNRB-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
PubChem CID 69070
IUPAC Name 2,6-dinitroaniline
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]

Specifications

Melting Point ∼136°C (decomposition)
Quantity 25 g
UN Number UN1596
Beilstein 2214886
Merck Index 14,3271
Solubility Information Insoluble in water.
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 2,6-Dinitroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.