missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylphenol, 97%, Thermo Scientific Chemicals

Product Code. 10450871
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-11-3
Molecular Formula C18H14O
Molecular Weight (g/mol) 246.31
MDL Number MFCD00009716
InChI Key ATGFTMUSEPZNJD-UHFFFAOYSA-N
Synonym 1,1':3',1-terphenyl-2'-ol, m-terphenyl-2'-ol, 2,6-diphenyl phenol, 2'-hydroxy-m-terphenyl, unii-la93w35m96, 2,6-di-phenylphenol, 2,6,-diphenylphenol, 2,6-di phenyl phenol, acmc-1ccsf
PubChem CID 75512

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 98°C to 103°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H4)2C6H3OH
Quantity 25 g
Beilstein 06, III, 3631
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol,acetone
Formula Weight 246.31
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 2, 6-Diphenylphenol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Diphenylphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.