missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylphenol, 98%, Thermo Scientific Chemicals

Product Code. 11303986
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-11-3
MDL Number MFCD00009716

Specifications

Melting Point 101°C to 103°C
Molecular Formula C5H2Br2O3
Quantity 25 g
Beilstein 2051224
Synonym 4,5-dibromo-2-furoic acid, 2,3-dibromofuran-5-carboxylic acid, 4,5-dibromo-2-furancarboxylic acid, 2-furancarboxylicacid, 4,5-dibromo, 2-furancarboxylic acid, 4,5-dibromo, zlchem 604, acmc-209xd1, ksc201e3j, 4,5-dibromo-2-furoicacid
InChI Key BHUVICYZDBUMIU-UHFFFAOYSA-N
SMILES C1=C(OC(=C1Br)Br)C(=O)O
IUPAC Name 4,5-dibromofuran-2-carboxylic acid
Molecular Weight (g/mol) 269.88
PubChem CID 4194832
Formula Weight 246.31
Percent Purity 98%
Chemical Name or Material 2,6-Diphenylphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.