missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Lutidine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
Description
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.
Solubility
Miscible with water.
Notes
Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.
Solubility
Miscible with water.
Notes
Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
Specifications
Specifications
Melting Point | -7°C |
Density | 0.923 |
Boiling Point | 143°C to 145°C |
Flash Point | 33°C (91°F) |
Odor | Unpleasant |
Refractive Index | 1.497 |
Quantity | 100 mL |
UN Number | UN1993 |
Beilstein | 105690 |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Product Identifier
- 2,6-Lutidine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 2,6-Lutidine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'