missing translation for 'onlineSavingsMsg'
Learn More
2-([7-(acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3,2-d][1,3]dioxin-8-yl]oxy)propanoic acid, Thermo Scientific™ Product Code.: 10681055

2-([7-(acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3,2-d][1,3]dioxin-8-yl]oxy)propanoic acid, Thermo Scientific™

Product Code. 10681055
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10681055

Brand: Thermo Scientific Maybridge RH01183CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 499104-69-7
Molecular Formula C25H29NO8
Molecular Weight (g/mol) 471.506
InChI Key JPPMVSNCFXDOJX-UHFFFAOYSA-N
Synonym 2-7-acetylamino-6-benzyloxy-2-phenylperhydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2r-2-4ar,6s,7r,8r,8as-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-6-benzyloxy-7-acetamido-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, benzyl n-acetyl-4,6-o-benzylidenemuramic acid, 2-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid
PubChem CID 3843768
IUPAC Name 2-[(7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl)oxy]propanoic acid
SMILES CC(C(=O)O)OC1C(C(OC2C1OC(OC2)C3=CC=CC=C3)OCC4=CC=CC=C4)NC(=O)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 471.5
Percent Purity 97%
Chemical Name or Material 2-{[7-(Acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3, 2-d][1, 3]dioxin-8-yl]oxy}propanoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-([7-(acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3,2-d][1,3]dioxin-8-yl]oxy)propanoic acid, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.