Learn More
MP Biomedicals™ 2-8 CELLsium™ Medium for Short-Term Cell Storage
2-8 CELLsius is a cytoprotective, protein free, ready to use bio-solution without DMSO for the short-term storage of cells or tissue.
Brand:  MP Biomedicals™ 092780354
 View more versions of this product
Product Code. 16309865
  • 808.00 € / 500mL
Explore more special offers
Description
Description
  • For short-term storage, add cold (2 to 8 °C) 2-8 CELLsium at a recommended cell concentration of 1 x 106 mL to 8 x 106 mL
  • The 2-8 CELLsium can be removed after centrifugation, cell concentration adjusted and transferred to an appropriate culture flask with growth medium
  • It will keep the cells healthy while in a temporary senescent phase, unlike regular growth media, which cannot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stored at 2 to 8°C
Sterile
500 mL
Liquid
Short-term cell storage