missing translation for 'onlineSavingsMsg'
Learn More
2,9-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals Product Code.: 15445879

2,9-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Product Code. 15445879
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15445879

Brand: Thermo Scientific Alfa Aesar J63698.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for the colorimetric determination of copper

2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-11-7
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00004973
InChI Key IYRGXJIJGHOCFS-UHFFFAOYSA-N
Synonym neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bvShow MoreShow Less
PubChem CID 65237
IUPAC Name 2,9-dimethyl-1,10-phenanthroline
SMILES CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C

Specifications

Melting Point 159°C to 164°C
Color Brown to White
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,6449
Solubility Information Slightly soluble in water and light petroleum; Soluble in methanol,ethanol,acetone,ether and benzene.
Formula Weight 208.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25I013 Certificate TypeCertificate of Analysis Date01/11/2022 Catalog NumberJ63698.14,J63698.06
Lot NumberY08H069 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ63698.14,J63698.06
Lot NumberC16Y024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63698.14,J63698.06
Lot NumberQ12E057 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63698.14,J63698.06
Lot NumberW25F022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63698.14,J63698.06
compliance-icons
Product Identifier
  • 2,9-Dimethyl-1,10-phenanthroline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,9-Dimethyl-1,10-phenanthroline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,9-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.