missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2-Acetamido-2-deoxy-D-glucopyranose, 98%
27.24 € - 588.93 €
Chemical Identifiers
CAS | 7512-17-6 |
---|---|
Molecular Formula | C8H15NO6 |
Molecular Weight (g/mol) | 221.21 |
MDL Number | MFCD00061615 |
InChI Key | OVRNDRQMDRJTHS-WZWLWIOANA-N |
Synonym | N-Acetyl-D-glucosamine |
IUPAC Name | N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10460521
|
Thermo Scientific Chemicals
270940100 |
10 g | Glass bottle |
27.24 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696784
|
Thermo Scientific Chemicals
270941000 |
100 g | Glass bottle |
155.78 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10063863
|
Thermo Scientific Chemicals
270945000 |
500 g | Plastic bottle |
588.93 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7512-17-6 | |
221.21 | |
OVRNDRQMDRJTHS-WZWLWIOANA-N | |
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
C8H15NO6 | |
MFCD00061615 | |
N-Acetyl-D-glucosamine | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Specifications
7512-17-6 | |
100.0 | |
2% max. (105°C, 4 hrs) | |
98% | |
MFCD00061615 | |
+ 42.00 (20.00°C c=2,water,2 hrs) | |
N-Acetyl-D-glucosamine | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O | |
221.21 | |
≥97.5% (HPLC) | |
2-Acetamido-2-deoxy-D-glucopyranose |
97.5 | |
White | |
Authentic | |
C8H15NO6 | |
15, 4494 | |
+39° to +45° (20°C, 589nm) (c=2, H2O, after 2 hrs) | |
OVRNDRQMDRJTHS-WZWLWIOANA-N | |
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
221.21 | |
Crystalline Powder |
RUO – Research Use Only