missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-6-chlorobenzoic acid, 99%, Thermo Scientific™

Product Code. 11342626
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19407-42-2
Molecular Formula C9H8ClNO3
Molecular Weight (g/mol) 213.617
MDL Number MFCD00051939
InChI Key VFHSJTHAMJFUCK-UHFFFAOYSA-N
Synonym 2-acetylamino-6-chlorobenzoic acid, 2-n-acetylamino-6-chlorobenzoic acid, 2-chloro-6-acetamidobenzoic acid, benzoic acid, 2-acetylamino-6-chloro, acmc-20ao6f, 2-acetamido-6-chloro-benzoic acid, 6-chloro-2-methylcarbonylamino benzoic acid
PubChem CID 88044
IUPAC Name 2-acetamido-6-chlorobenzoic acid
SMILES CC(=O)NC1=C(C(=CC=C1)Cl)C(=O)O

Specifications

Melting Point 216°C to 218°C
Quantity 25 g
Beilstein 784450
Formula Weight 213.62
Percent Purity 99%
Chemical Name or Material 2-Acetamido-6-chlorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'