missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxyisobutyryl bromide, 96%, Thermo Scientific Chemicals

Product Code. 11426780
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40635-67-4
Molecular Formula C6H9BrO3
Molecular Weight (g/mol) 209.039
MDL Number MFCD00040923
InChI Key OOKAXSHFTDPZHP-UHFFFAOYSA-N
Synonym 2-acetoxyisobutyryl bromide, 2-acetoxy-2-methylpropionyl bromide, 1-bromocarbonyl-1-methylethyl acetate, alpha-acetoxy-isobutyryl bromide, a-acetoxyisobutyryl bromide, alpha-acetoxyisobutyryl bromide, 2-acetoxyisobutyryl bromide;2-acetoxy-2-methylpropionyl bromide, propanoyl bromide, 2-acetyloxy-2-methyl, aibb, pubchem17437
PubChem CID 2724715
IUPAC Name (1-bromo-2-methyl-1-oxopropan-2-yl) acetate
SMILES CC(=O)OC(C)(C)C(=O)Br

Specifications

Density 1.42
Boiling Point 100°C to 110°C (25 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.457
Quantity 5 g
UN Number UN3265
Beilstein 2432585
Sensitivity Moisture sensitive
Formula Weight 209.05
Percent Purity 96%
Chemical Name or Material 2-Acetoxyisobutyryl bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.