missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-methylpyrrole, 98%, Thermo Scientific Chemicals

Product Code. 11418286
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-16-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00003089
InChI Key NZFLWVDXYUGFAV-UHFFFAOYSA-N
Synonym 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole
PubChem CID 61240
ChEBI CHEBI:59982
IUPAC Name 1-(1-methylpyrrol-2-yl)ethanone
SMILES CN1C=CC=C1C(C)=O

Specifications

Density 1.04
Boiling Point 200°C to 202°C
Flash Point 68°C (154°F)
Odor Pleasant
Refractive Index 1.542
Quantity 100 g
Beilstein 111887
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Acetyl-1-methylpyrrole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'