missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3-bromothiophene, 97%, Thermo Scientific™

Product Code. 11364556
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42877-08-7
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD00794287
InChI Key ZUXPELAHJQSZTE-UHFFFAOYSA-N
Synonym 2-acetyl-3-bromothiophene, 1-3-bromothiophen-2-yl ethanone, 1-3-bromothiophen-2-yl ethan-1-one, 1-3-bromo-2-thienyl-1-ethanone, bromothienylethanone, 1-3-bromothien-2-yl ethanone, 1-3-bromo-2-thienyl ethanone, pubchem7280, acmc-20ai7w
PubChem CID 2764496
SMILES CC(=O)C1=C(Br)C=CS1

Specifications

Melting Point 41°C to 45°C
Boiling Point 102°C to 105°C (4 mmHg)
Refractive Index 1.614
Quantity 5 g
Beilstein 116973
Sensitivity Light sensitive
IUPAC Name 1-(3-bromothiophen-2-yl)ethan-1-one
Formula Weight 205.07
Percent Purity 97%
Chemical Name or Material 2-Acetyl-3-bromothiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'