missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals

Product Code. 11489166
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3900-45-6
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00021643
InChI Key GGWCZBGAIGGTDA-UHFFFAOYSA-N
Synonym 2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
PubChem CID 77506
IUPAC Name 1-(6-methoxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 107°C to 109°C
Boiling Point 155°C to 160°C (0.4 mmHg)
Quantity 25 g
Beilstein 1871098
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 2-Acetyl-6-methoxynaphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'