missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific™

Product Code. 15499015
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81885-67-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD10016667
InChI Key XWVFMJLNNGXNSG-UHFFFAOYSA-N
Synonym 1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethanone, 2-acetyl-1,2,3,4-tetrahydroisoquinolin-7-amine, 2-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 7-isoquinolinamine, 2-acetyl-1,2,3,4-tetrahydro, 1-7-amino-3,4-dihydro-1h-isoquinolin-2-yl ethanone, n-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl ethanone, 1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethan-1-one, ethanone, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl, 1-7-amino-1,2,3,4-tetrahydroisoquinolin-2-yl ethan-1-one
PubChem CID 12811352
IUPAC Name 1-(7-amino-3,4-dihydro-1H-isoquinolin-2-yl)ethanone
SMILES CC(=O)N1CCC2=C(C1)C=C(C=C2)N

Specifications

Melting Point 107°C to 111°C
Color Brown
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 190.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.