missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzo[b]furan, 99%, Thermo Scientific Chemicals

42.30 € - 154.00 €

Chemical Identifiers

CAS 1646-26-0
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00005849
InChI Key YUTFQTAITWWGFH-UHFFFAOYSA-N
Synonym 2-acetylbenzofuran, 1-benzofuran-2-yl ethanone, ethanone, 1-2-benzofuranyl, benzofuran-2-yl methyl ketone, 2-acetylcoumarone, 2-acetylcumarone, 2-benzofuranyl methyl ketone, benzo b furan-2-yl methyl ketone, 1-2-benzofuranyl ethanone, 2-acetylbenzo b furan
PubChem CID 15435
SMILES CC(=O)C1=CC2=CC=CC=C2O1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323237
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13771.06
5 g
42.30 €
5g
Please to purchase this item. Need a web account? Register with us today!
11333237
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13771.14
25 g
154.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

1646-26-0
160.17
YUTFQTAITWWGFH-UHFFFAOYSA-N
15435
C10H8O2
MFCD00005849
2-acetylbenzofuran, 1-benzofuran-2-yl ethanone, ethanone, 1-2-benzofuranyl, benzofuran-2-yl methyl ketone, 2-acetylcoumarone, 2-acetylcumarone, 2-benzofuranyl methyl ketone, benzo b furan-2-yl methyl ketone, 1-2-benzofuranyl ethanone, 2-acetylbenzo b furan
CC(=O)C1=CC2=CC=CC=C2O1

Specifications

1646-26-0
110°C to 113°C (3 mmHg)
C10H8O2
117910
Insoluble in water.
CC(=O)C1=CC2=CC=CC=C2O1
160.17
160.17
2-Acetylbenzo[b]furan
70°C to 72°C
>110°C (230°F)
MFCD00005849
2-acetylbenzofuran, 1-benzofuran-2-yl ethanone, ethanone, 1-2-benzofuranyl, benzofuran-2-yl methyl ketone, 2-acetylcoumarone, 2-acetylcumarone, 2-benzofuranyl methyl ketone, benzo b furan-2-yl methyl ketone, 1-2-benzofuranyl ethanone, 2-acetylbenzo b furan
YUTFQTAITWWGFH-UHFFFAOYSA-N
1-(1-benzofuran-2-yl)ethan-1-one
15435
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 216-706-9

missing translation for 'rtecsNumber' : OB1203000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only