missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylnaphthalene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Description
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.
Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.
Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Specifications
Melting Point | 52°C to 56°C |
Boiling Point | 300°C to 301°C |
Flash Point | 168°C (334°F) |
Quantity | 100 g |
UN Number | UN3077 |
Beilstein | 774965 |
Solubility Information | Soluble in ethanol,acetone and ether. Insoluble in water. |
Formula Weight | 170.21 |
Percent Purity | 99% |
Chemical Name or Material | 2-Acetylnaphthalene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'