missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylphenylboronic acid, 96%, Thermo Scientific Chemicals

Product Code. 10487582
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308103-40-4
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01321263
InChI Key ZKAOVABYLXQUTI-UHFFFAOYSA-N
Synonym 2-acetylphenyl boronic acid, boronic acid, 2-acetylphenyl, 2'-boronoacetophenone, 1-2-boronophenyl ethan-1-one, 2-acetylbenzeneboronic acid, 2-acetylbenzenboronicacid, 1-2-dihydroxyboranyl phenyl ethan-1-one, 2-acetylphenylboronicacid, pubchem5373, acmc-1agh6
PubChem CID 2734309
IUPAC Name (2-acetylphenyl)boronic acid
SMILES CC(=O)C1=CC=CC=C1B(O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 170.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.97
Percent Purity 96%
Physical Form Chunks and/or Powder
Chemical Name or Material 2-Acetylphenylboronic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.