Learn More
2-Adamantanone, 98%, Thermo Scientific Chemicals
35.50 € - 288.00 €
Chemical Identifiers
CAS | 700-58-3 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.221 |
MDL Number | MFCD00074737 |
InChI Key | IYKFYARMMIESOX-UHFFFAOYSA-N |
Synonym | 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one |
PubChem CID | 64151 |
ChEBI | CHEBI:40611 |
IUPAC Name | adamantan-2-one |
SMILES | C1C2CC3CC1CC(C2)C3=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482427
|
Thermo Scientific Alfa Aesar
A14275.06 |
5 g |
35.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492427
|
Thermo Scientific Alfa Aesar
A14275.14 |
25 g |
102.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402437
|
Thermo Scientific Alfa Aesar
A14275.22 |
100 g |
288.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (±)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (+/-)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
700-58-3 | |
150.221 | |
IYKFYARMMIESOX-UHFFFAOYSA-N | |
64151 | |
adamantan-2-one |
C10H14O | |
MFCD00074737 | |
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one | |
CHEBI:40611 | |
C1C2CC3CC1CC(C2)C3=O |
Specifications
700-58-3 | |
C10H14O | |
1210235 | |
IYKFYARMMIESOX-UHFFFAOYSA-N | |
adamantan-2-one | |
64151 | |
150.22 | |
2-Adamantanone |
∼257°C (sublimation) | |
MFCD00074737 | |
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one | |
C1C2CC3CC1CC(C2)C3=O | |
150.221 | |
CHEBI:40611 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 211-847-2
missing translation for 'rtecsNumber' : AU5018000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only