missing translation for 'onlineSavingsMsg'
Learn More
2-Allylphenol, 98+%, Thermo Scientific Chemicals Product Code.: 11439642

2-Allylphenol, 98+%, Thermo Scientific Chemicals

Product Code. 11439642
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11439642

Brand: Thermo Scientific Alfa Aesar A10755.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

Solubility
Soluble in water(7 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1745-81-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002250
InChI Key QIRNGVVZBINFMX-UHFFFAOYSA-N
Synonym 2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenolShow MoreShow Less
PubChem CID 15624
ChEBI CHEBI:39826
IUPAC Name 2-prop-2-enylphenol
SMILES C=CCC1=CC=CC=C1O

Specifications

Melting Point -6°C
Density 1.024
Boiling Point 220°C to 222°C
Flash Point 88°C (190°F)
Odor Phenol-like
Refractive Index 1.545
Quantity 100 g
UN Number UN2922
Beilstein 742121
Solubility Information Soluble in water(7g/L).
Formula Weight 134.18
Percent Purity ≥98%
Chemical Name or Material 2-Allylphenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number28118 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10755.22,A10755.36
Lot Number10157008 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10755.22,A10755.36
Lot Number10162301 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10755.22,A10755.36
Lot Number10175036 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10755.22,A10755.36
Lot NumberF2710A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10755.22,A10755.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Allylphenol, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.