Learn More
2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals
A useful biological buffer
27.34 € - 163.82 €
Chemical Identifiers
CAS | 115-69-5 |
---|---|
Molecular Formula | C4H11NO2 |
Molecular Weight (g/mol) | 105.137 |
MDL Number | MFCD00004678 |
InChI Key | UXFQFBNBSPQBJW-UHFFFAOYSA-N |
Synonym | 2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol |
PubChem CID | 1531 |
ChEBI | CHEBI:991 |
IUPAC Name | 2-amino-2-methylpropane-1,3-diol |
SMILES | CC(CO)(CO)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15475409
|
Thermo Scientific Alfa Aesar
J63144.14 |
25 g |
27.34 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15485409
|
Thermo Scientific Alfa Aesar
J63144.22 |
100 g |
75.27 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15495409
|
Thermo Scientific Alfa Aesar
J63144.30 |
250 g |
163.82 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.
Solubility
Soluble in water and other organic solvents.
Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, aluminum, copper, brass and strong acids.
Chemical Identifiers
115-69-5 | |
105.137 | |
UXFQFBNBSPQBJW-UHFFFAOYSA-N | |
1531 | |
2-amino-2-methylpropane-1,3-diol |
C4H11NO2 | |
MFCD00004678 | |
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol | |
CHEBI:991 | |
CC(CO)(CO)N |
Specifications
115-69-5 | |
White | |
C4H11NO2 | |
MFCD00004678 | |
14,448 | |
Soluble in water and other organic solvents. | |
CC(CO)(CO)N | |
105.137 | |
CHEBI:991 | |
≥99% | |
2-Amino-2-methyl-1,3-propanediol |
100°C to 110°C | |
151°C (10 mmHg) | |
(HOCH2)2C(NH2)CH3 | |
635708 | |
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol | |
UXFQFBNBSPQBJW-UHFFFAOYSA-N | |
2-amino-2-methylpropane-1,3-diol | |
1531 | |
105.14 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P390-P501c
H290-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 204-100-7
missing translation for 'rtecsNumber' : TY2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2-Amino-2-methyl-1,3-propanediol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : 2-Amino-2-methyl-1,3-propanediol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.