missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water, Thermo Scientific Chemicals

Product Code. 11349699
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

Solubility
Immiscible with water. Miscible with alcohols.

Notes
Incompatible withoxidizing agents, strong acids, copper, brass and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(N)CO

Specifications

Melting Point 24°C to 28°C
Density 0.934
pH 11.3
Boiling Point 164°C to 166°C
Flash Point 67°C (152°F)
Odor Amine-like
Refractive Index 1.4455
Quantity 100 mL
Beilstein 505979
Merck Index 14,449
Solubility Information Immiscible with water. Miscible with alcohols.
Formula Weight 89.14
Percent Purity 95%
Assay may cont. ≈5% water
Chemical Name or Material 2-Amino-2-methyl-1-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'