missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-2-thioxoethyl pivalate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | (CH3)3CCO2CH2C(S)NH2 |
Quantity | 1 g |
Formula Weight | 175.25 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-2-thioxoethyl pivalate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Amino-2-thioxoethyl pivalate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction