missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. 11313406
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, bases and sodium hypochlorite.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3032-32-4
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00052390
InChI Key MMIREQFMYZQWLU-UHFFFAOYSA-N
PubChem CID 604542
IUPAC Name 2-amino-3,6-dichlorobenzoic acid
SMILES NC1=C(Cl)C=CC(Cl)=C1C(O)=O

Specifications

Melting Point 150°C to 154°C
Quantity 1 g
Beilstein 3266879
Synonym benzoic acid, 2-amino-3,6-dichloro, 3,6-dichloroanthranilic acid, 2-amino-3,6-dichlorobenzoicacid, acmc-1afqi, 6-amino-2,5-dichlorobenzoic acid, benzoic acid,2-amino-3,6-dichloro, 2-azanyl-3,6-bis chloranyl benzoic acid
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 95%
Chemical Name or Material 2-Amino-3,6-dichlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'