Learn More
2-Amino-3-nitrophenol, 98%, Thermo Scientific Chemicals
62.70 € - 190.00 €
Chemical Identifiers
CAS | 603-85-0 |
---|---|
Molecular Formula | C6H6N2O3 |
Molecular Weight (g/mol) | 154.13 |
MDL Number | MFCD00010875 |
InChI Key | KUCWUAFNGCMZDB-UHFFFAOYSA-N |
Synonym | phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol |
PubChem CID | 4115495 |
IUPAC Name | 2-amino-3-nitrophenol |
SMILES | NC1=C(O)C=CC=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10250353
|
Thermo Scientific Acros
382040050 |
5 g | Glass bottle |
62.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10075932
|
Thermo Scientific Acros
382040250 |
25 g | Glass bottle |
190.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
603-85-0 | |
154.13 | |
KUCWUAFNGCMZDB-UHFFFAOYSA-N | |
4115495 | |
NC1=C(O)C=CC=C1[N+]([O-])=O |
C6H6N2O3 | |
MFCD00010875 | |
phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol | |
2-amino-3-nitrophenol |
Specifications
603-85-0 | |
100.0 | |
Red to Brown | |
97.5% min. (HPLC) | |
H2NC6H3(NO2)OH | |
phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol | |
KUCWUAFNGCMZDB-UHFFFAOYSA-N | |
2-amino-3-nitrophenol | |
4115495 | |
98% | |
2-Amino-3-nitrophenol |
97.5 | |
215.0°C | |
Authentic | |
C6H6N2O3 | |
MFCD00010875 | |
Solubility in water: slightly soluble. | |
NC1=C(O)C=CC=C1[N+]([O-])=O | |
154.13 | |
154.12 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-060-1
- 2-Amino-3-nitrophenol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.