missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals

Product Code. 15464798
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 30235-26-8
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.225
MDL Number MFCD00460417
InChI Key BLKHMTAXNXLDJP-UHFFFAOYSA-N
Synonym 4-pyridin-2-yl thiazol-2-amine, 4-pyridin-2-yl-thiazol-2-ylamine, 4-pyridin-2-yl-1,3-thiazol-2-amine, 2-amino-4-2-pyridyl thiazole, 2-thiazolamine, 4-2-pyridinyl, 4-2-pyridinyl-1,3-thiazol-2-amine, 4-2-pyridinyl-2-thiazolamine, 4-2-pyridinyl thiazol-2-amine, 4-2-pyridyl-1,3-thiazole-2-ylamine, 4-pyridin-2-yl-thiazole-2-ylamine
PubChem CID 1092459
IUPAC Name 4-pyridin-2-yl-1,3-thiazol-2-amine
SMILES C1=CC=NC(=C1)C2=CSC(=N2)N

Spezifikation

Color Brown
Quantity 2 g
Formula Weight 177.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(2-pyridyl)thiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'