missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, Thermo Scientific™

Product Code. 10621883
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2192-06-5
Molecular Formula C3H4ClN2OS
Molecular Weight (g/mol) 151.59
InChI Key CJPDNRMSRPXGQO-UHFFFAOYSA-N
Synonym 2-aminothiazol-4 5h-one hydrochloride, 2-aminothiazolinone hcl, 2-amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, 2-aminothiazolinone hydrochloride, 2-amino-1,3-thiazol-4 5h-one hydrochloride, 4 5h-thiazolone, 2-amino-hydrochloride, 2-amino-1,3-thiazolin-4-one, chloride, 2-aminothiazole-4-one hydrochloride, 2-amino-1,3-thiazol-4-one hydrochloride, c3h4n2os.hcl
PubChem CID 2782348
IUPAC Name 2-amino-1,3-thiazol-4-one;hydrochloride
SMILES C1C(=O)N=C(S1)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 152.6
Percent Purity 97%
Chemical Name or Material 2-Amino-4, 5-dihydro-1, 3-thiazol-4-one hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.