missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 15446318

2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15446318
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15446318

Brand: Thermo Scientific Alfa Aesar H59878.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71574-33-9
Molecular Formula C5H9ClN2S
Molecular Weight (g/mol) 164.651
MDL Number MFCD00012711
InChI Key XYTUTNQRQLAZLK-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazol-2-amine hydrochloride, 2-amino-4,5-dimethylthiazole hydrochloride, 4,5-dimethyl-1,3-thiazol-2-amine hydrochloride, dimethyl-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 4,5-dimethyl-, monohydrochloride, acmc-209oj8, ksc496i8r, 4,5-dimethylthiazol-2-amine hcl salt, 4,5-dimethylthiazol-2-amine hcl, 2-amino-4,5-dimethyl thiazole hydrochloride
PubChem CID 16211387
IUPAC Name 4,5-dimethyl-1,3-thiazol-2-amine;hydrochloride
SMILES CC1=C(SC(=N1)N)C.Cl

Specifications

Melting Point 273°C to 274°C (decomposition)
Quantity 50 g
Sensitivity Hygroscopic
Formula Weight 164.66
Percent Purity 98%
Chemical Name or Material 2-Amino-4,5-dimethylthiazole hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.