missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-diphenylthiazole, 97%, Thermo Scientific™

Product Code. p-7034157
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15498277

Brand: Thermo Scientific Alfa Aesar H51818.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne).

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6318-74-7
Molecular Formula C15H12N2S
Molecular Weight (g/mol) 252.335
MDL Number MFCD00438803
InChI Key XDWGEGZDMFHZBL-UHFFFAOYSA-N
PubChem CID 73222
IUPAC Name 4,5-diphenyl-1,3-thiazol-2-amine
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)N)C3=CC=CC=C3

Specifications

Odor Odorless
Quantity 1 g
Synonym 2-amino-4,5-diphenylthiazole, 2-thiazolamine, 4,5-diphenyl, 4,5-diphenyl-thiazol-2-ylamine, 4,5-diphenylthiazol-2-amine, diphenyl-1,3-thiazol-2-amine, 2-thiazolamine,4,5-diphenyl, thiazole, 2-amino-4,5-diphenyl, 4,5-diphenyl-2-thiazolamine, monohydrochloride, 4,5-diphenyl-1,3-thiazole-2-ylamine, ccris 8105
Solubility Information Insoluble in water. Soluble in chloroform and methanol.
Formula Weight 252.33
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5-diphenylthiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.