missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromophenol, 98%, Thermo Scientific™

22.70 € - 205.00 €

Chemical Identifiers

CAS 40925-68-6
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD00235171
InChI Key JHRIPENGTGSNPJ-UHFFFAOYSA-N
Synonym phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p
PubChem CID 351840
IUPAC Name 2-amino-4-bromophenol
SMILES NC1=CC(Br)=CC=C1O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15565246
View Documents
Thermo Scientific Acros
457480010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15575246
View Documents
Thermo Scientific Acros
457480050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15585246
View Documents
Thermo Scientific Acros
457480250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

40925-68-6
188.02
JHRIPENGTGSNPJ-UHFFFAOYSA-N
351840
NC1=CC(Br)=CC=C1O
C6H6BrNO
MFCD00235171
phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p
2-amino-4-bromophenol

Specifications

40925-68-6
97.5% min. (HPLC)
C6H6BrNO
phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p
NC1=CC(Br)=CC=C1O
188.02
188.02
2-Amino-4-bromophenol
Authentic
Glass Bottle
MFCD00235171
JHRIPENGTGSNPJ-UHFFFAOYSA-N
2-amino-4-bromophenol
351840
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

Product Identifier
  • 2-Amino-4-bromophenol, 98%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.