missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzoic acid, 98%

Product Code. 11389689
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
5g
This item is not returnable. View return policy

Product Code. 11389689

Brand: Thermo Scientific Alfa Aesar A10201.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-77-0 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-77-0
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007778
InChI Key JYYLQSCZISREGY-UHFFFAOYSA-N
Synonym 4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
PubChem CID 66646
IUPAC Name 2-amino-4-chlorobenzoic acid
SMILES NC1=CC(Cl)=CC=C1C(O)=O

Specifications

Melting Point ∼232°C (decomposition)
Quantity 100 g
Beilstein 743349
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-4-chlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.