missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-4-chlorothiophenol, 96%, Thermo Scientific Chemicals Product Code.: 11471657

2-Amino-4-chlorothiophenol, 96%, Thermo Scientific Chemicals

Product Code. 11471657
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11471657

Brand: Thermo Scientific Alfa Aesar B25640.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-00-8
Molecular Formula C6H6ClNS
Molecular Weight (g/mol) 159.631
MDL Number MFCD00792528
InChI Key NGIRMPARLVGMPX-UHFFFAOYSA-N
Synonym 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
PubChem CID 12000
IUPAC Name 2-amino-4-chlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)N)S

Specifications

Melting Point 52°C to 59°C
Odor Unpleasant
Quantity 5 g
UN Number UN1759
Sensitivity Air Sensitive
Formula Weight 159.64
Percent Purity 96%
Chemical Name or Material 2-Amino-4-chlorothiophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-4-chlorothiophenol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.