missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorothiophenol, 96%, Thermo Scientific™

Product Code. 13448678
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-00-8
Molecular Formula C6H6ClNS
Molecular Weight (g/mol) 159.64
InChI Key NGIRMPARLVGMPX-UHFFFAOYSA-N
Synonym 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
PubChem CID 12000
IUPAC Name 2-amino-4-chlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)N)S

Specifications

Melting Point 56.0°C to 61.0°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(Cl)SH
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 159.64
Percent Purity 96%
Chemical Name or Material 2-Amino-4-chlorothiophenol
compliance-icons
Product Identifier
  • 2-Amino-4-chlorothiophenol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'