missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-imino-2-thiazoline hydrochloride, 99%, Thermo Scientific Chemicals
36.00 € - 111.00 €
Chemical Identifiers
CAS | 36518-76-0 |
---|---|
Molecular Formula | C3H6ClN3S |
Molecular Weight (g/mol) | 151.612 |
MDL Number | MFCD00012721 |
InChI Key | ANDLBJXZBLTCDP-UHFFFAOYSA-N |
Synonym | 2-amino-4-imino-2-thiazoline hydrochloride, 2,4-diiminothiazolidine hydrochloride, 2-imino-2,5-dihydrothiazol-4-amine hydrochloride, 2-imino-5h-1,3-thiazol-4-amine hydrochloride, 4-imino-5h-1,3-thiazol-2-amine hydrochloride, acmc-1aeug, 4-imino-1,3-thiazoline-2-ylamine, chloride, 4-imino-4,5-dihydrothiazol-2-amine hydrochloride |
PubChem CID | 2723857 |
IUPAC Name | 2-imino-5H-1,3-thiazol-4-amine;hydrochloride |
SMILES | C1C(=NC(=N)S1)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387605
|
Thermo Scientific Alfa Aesar
L05844.06 |
5 g |
36.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397605
|
Thermo Scientific Alfa Aesar
L05844.14 |
25 g |
111.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
36518-76-0 | |
151.612 | |
ANDLBJXZBLTCDP-UHFFFAOYSA-N | |
2723857 | |
C1C(=NC(=N)S1)N.Cl |
C3H6ClN3S | |
MFCD00012721 | |
2-amino-4-imino-2-thiazoline hydrochloride, 2,4-diiminothiazolidine hydrochloride, 2-imino-2,5-dihydrothiazol-4-amine hydrochloride, 2-imino-5h-1,3-thiazol-4-amine hydrochloride, 4-imino-5h-1,3-thiazol-2-amine hydrochloride, acmc-1aeug, 4-imino-1,3-thiazoline-2-ylamine, chloride, 4-imino-4,5-dihydrothiazol-2-amine hydrochloride | |
2-imino-5H-1,3-thiazol-4-amine;hydrochloride |
Specifications
36518-76-0 | |
C3H6ClN3S | |
5109728 | |
ANDLBJXZBLTCDP-UHFFFAOYSA-N | |
2-imino-5H-1,3-thiazol-4-amine;hydrochloride | |
2723857 | |
99% |
>300°C | |
MFCD00012721 | |
2-amino-4-imino-2-thiazoline hydrochloride, 2,4-diiminothiazolidine hydrochloride, 2-imino-2,5-dihydrothiazol-4-amine hydrochloride, 2-imino-5h-1,3-thiazol-4-amine hydrochloride, 4-imino-5h-1,3-thiazol-2-amine hydrochloride, acmc-1aeug, 4-imino-1,3-thiazoline-2-ylamine, chloride, 4-imino-4,5-dihydrothiazol-2-amine hydrochloride | |
C1C(=NC(=N)S1)N.Cl | |
151.612 | |
151.62 | |
2-Amino-4-imino-2-thiazoline hydrochloride |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 253-083-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only