missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-methylbenzoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 151.16 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-4-methylbenzoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction