missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzoic acid, 95%, Thermo Scientific™

Product Code. 10445912
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2305-36-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00047853
InChI Key RPGKFFKUTVJVPY-UHFFFAOYSA-N
Synonym 4-methylanthranilic acid, benzoic acid, 2-amino-4-methyl, 2-amino-4-methyl-benzoic acid, 2-amino-p-toluic acid, 3-amino-4-carboxytoluene, 2-carboxy-5-methylaniline, p-toluic acid, 2-amino, rarechem al bo 0820, benzoic acid, 2-amino-4-methyl-9ci, 2-azanyl-4-methyl-benzoic acid
PubChem CID 75316
IUPAC Name 2-amino-4-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)N

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.