missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 10319360
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-88-7
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007823
InChI Key CUKXRHLWPSBCTI-UHFFFAOYSA-N
Synonym 5-bromoanthranilic acid, benzoic acid, 2-amino-5-bromo, anthranilic acid, 5-bromo, 2-amino-5-bromo-benzoic acid, 5-bromo-2-aminobenzoic acid, 2-amino-5-bromobenzoicacid, 2-amino-5-bromo benzoic acid, 5-bromo anthranilic acid, buttpark 89\07-49, 5-bromo-2-amino-benzoic acid
PubChem CID 79858
IUPAC Name 2-amino-5-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 218°C to 220°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(Br)CO2H
Quantity 5 g
Merck Index 15, 1412
Formula Weight 216.03
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-bromobenzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'