missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals Product Code.: 10020700

2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals

Product Code. 10020700
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10020700

Brand: Thermo Scientific Acros 103380050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-80-3
Molecular Formula C7H5ClN2O
Molecular Weight (g/mol) 168.58
MDL Number MFCD00005770
InChI Key YGCODSQDUUUKIV-UHFFFAOYSA-N
Synonym zoxazolamine, 2-amino-5-chlorobenzoxazole, 5-chlorobenzo d oxazol-2-amine, flexin, 2-benzoxazolamine, 5-chloro, contrazole, deflexol, flexilon, zoxamin, zoxine
PubChem CID 6103
ChEBI CHEBI:35053
IUPAC Name 5-chloro-1,3-benzoxazol-2-amine
SMILES NC1=NC2=CC(Cl)=CC=C2O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180.0°C to 185.0°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10399
Formula Weight 168.58
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-chlorobenzoxazole, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-chlorobenzoxazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.