missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

245.00 € - 872.00 €

Chemical Identifiers

CAS 1073371-77-3
Molecular Formula C12H17BClNO2
Molecular Weight (g/mol) 253.53
MDL Number MFCD06795672
InChI Key GVJZHGCVSYBPIM-UHFFFAOYSA-N
Synonym 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid, pinacol ester, 2-amino-5-chlorophenylboronic acid pinacol ester, 4-chloro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid,pinacol ester, 2-amino-5-chlorobenzeneboronic acid, pinacol ester, 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
PubChem CID 17750205
IUPAC Name 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11996031
View Documents Promotion Details
Thermo Scientific Acros
444470010
1 g
245.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11906041
View Documents Promotion Details
Thermo Scientific Acros
444470050
5 g
872.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1073371-77-3
253.53
GVJZHGCVSYBPIM-UHFFFAOYSA-N
17750205
CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1
C12H17BClNO2
MFCD06795672
4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid, pinacol ester, 2-amino-5-chlorophenylboronic acid pinacol ester, 4-chloro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid,pinacol ester, 2-amino-5-chlorobenzeneboronic acid, pinacol ester, 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

Specifications

1073371-77-3
96% min. (GC)
C12H17BClNO2
4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid, pinacol ester, 2-amino-5-chlorophenylboronic acid pinacol ester, 4-chloro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-chlorophenylboronic acid,pinacol ester, 2-amino-5-chlorobenzeneboronic acid, pinacol ester, 4-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1
253.53
253.54
2-Amino-5-chlorophenylboronic acid pinacol ester
91°C to 93°C
Glass Bottle
MFCD06795672
GVJZHGCVSYBPIM-UHFFFAOYSA-N
4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
17750205
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning

Product Identifier
  • 2-Amino-5-chlorophenylboronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only