missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11446974
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-08-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00055566
InChI Key FPQMGQZTBWIHDN-UHFFFAOYSA-N
Synonym 5-fluoroanthranilic acid, 2-amino-5-fluoro-benzoic acid, benzoic acid, 2-amino-5-fluoro, 5-faa, 5-fluoro-2-aminobenzoic acid, 4own, 5rg, pubchem1322, 5-fluoroanthanilic acid, acmc-209jyo
PubChem CID 101412
ChEBI CHEBI:78042
IUPAC Name 2-amino-5-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)N

Specifications

Melting Point 182°C to 184°C
Quantity 25 g
Beilstein 2803664
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-fluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'